Gene Transcripts
- Gene Symbol
- C1orf116
- Entrez Gene ID
- 79098
- Gene Name
- chromosome 1 open reading frame 116
- Gene Aliases
- SARG
- Chromosome Location
- Chr. 1: 207018521 - 207032761 on Build GRCh38
- UniGene ID
- Hs.32417
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BG150416.1 |
- |
1 |
57 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AF007835.1 |
- |
1 |
173 |
|
Chr. 1 - 207018401 - 207018900 [+] |
View Full Product Details