Gene Transcripts

Gene Symbol
ZNF75D
Entrez Gene ID
7626
Gene Name
zinc finger protein 75D
Gene Aliases
D8C6, ZKSCAN24, ZNF75, ZNF82, ZSCAN28
Chromosome Location
Chr. X: 135248589 - 135344109 on Build GRCh38
UniGene ID
Hs.371717
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_110381.1 3 1202
GenBank mRNA AY730281.1 - 2 395
BC029787.1 - 3 1202
BG186944.1 - 2 221

View Full Product Details