Gene Transcripts
- Gene Symbol
- SLC25A29
- Entrez Gene ID
- 123096
- Gene Name
- solute carrier family 25 member 29
- Gene Aliases
- C14orf69, CACL, ORNT3
- Chromosome Location
- Chr. 14: 100277986 - 100306547 on Build GRCh38
- UniGene ID
- Hs.578109
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
R33710.1 |
- |
|
110 |
|
Chr. - - [-] |
GenBankmRNA |
AK094455.1 |
- |
|
950 |
|
Chr. - - [-] |
GenBankmRNA |
DA328919.1 |
- |
1 |
204 |
|
Chr. 14 - 100296440 - 100297431 [-] |
GenBankmRNA |
DA328937.1 |
- |
1 |
204 |
|
Chr. 14 - 100296457 - 100297431 [-] |
View Full Product Details