Gene Transcripts
- Gene Symbol
- ST5
- Entrez Gene ID
- 6764
- Gene Name
- suppression of tumorigenicity 5
- Gene Aliases
- DENND2B, HTS1, p126
- Chromosome Location
- Chr. 11: 8693352 - 8910951 on Build GRCh38
- UniGene ID
- Hs.117715
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BX647474.1 |
- |
2 |
3650 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AK123380.1 |
- |
1 |
2011 |
|
Chr. 11 - 8806934 - 8809859 [-] |
View Full Product Details