Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004163.4 | NP_004154.2 | 6 | 5358 | |
XM_005266733.1 | XP_005266790.1 | 7 | 5279 | ||
XM_006722518.1 | XP_006722581.1 | 6 | 1868 | ||
XM_017025913.1 | XP_016881402.1 | 9 | 6112 | ||
XM_017025914.1 | XP_016881403.1 | 8 | 5507 | ||
GenBank mRNA | AF131784.1 | - | 1 | 925 |