Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004163.4 | NP_004154.2 | 6 | 6555 | |
XM_005266733.1 | XP_005266790.1 | 7 | 6476 | ||
XM_006722518.1 | XP_006722581.1 | 6 | 3065 | ||
XM_011526131.1 | XP_011524433.1 | 7 | 1642 | ||
XM_017025913.1 | XP_016881402.1 | 9 | 7309 | ||
XM_017025914.1 | XP_016881403.1 | 8 | 6704 | ||
GenBank mRNA | AF131784.1 | - | 1 | 2122 |