Gene Transcripts

Gene Symbol
RAB27B
Entrez Gene ID
5874
Gene Name
RAB27B, member RAS oncogene family
Gene Aliases
C25KG
Chromosome Location
Chr. 18: 54717842 - 54895516 on Build GRCh38
UniGene ID
Hs.25318
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004163.4 NP_004154.2 6 5076
XM_005266733.1 XP_005266790.1 7 4997
XM_006722518.1 XP_006722581.1 6 1586
XM_017025913.1 XP_016881402.1 9 5830
XM_017025914.1 XP_016881403.1 8 5225
GenBank mRNA AF131784.1 - 1 643

View Full Product Details