Gene Transcripts

Gene Symbol
LRRC28
Entrez Gene ID
123355
Gene Name
leucine rich repeat containing 28
Gene Aliases
Chromosome Location
Chr. 15: 99251362 - 99390732 on Build GRCh38
UniGene ID
Hs.578684
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001284400.2 NP_001271329.1 9 3575
NM_001321675.1 NP_001308604.1 10 3731
NM_001321676.1 NP_001308605.1 11 3723
NM_001321677.1 NP_001308606.1 9 3528
NM_001321678.1 NP_001308607.1 9 3423
NM_001321679.1 NP_001308608.1 9 3694
NM_001321680.1 NP_001308609.1 9 3690
NM_144598.4 NP_653199.2 10 3735
NR_135753.1 10 3640
NR_135754.1 10 3488
NR_135755.1 9 3595
NR_135756.1 9 3599
NR_135757.1 11 3709
NR_135758.1 10 3763
NR_135759.1 8 3483


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA CR593703.1 - 1 883 Chr. 15 - 99387611 - 99389069 [+]
GenBankmRNA BE782162.1 - 125 Chr. - - [-]

View Full Product Details