Gene Transcripts
- Gene Symbol
- LRRC28
- Entrez Gene ID
- 123355
- Gene Name
- leucine rich repeat containing 28
- Gene Aliases
- Chromosome Location
- Chr. 15: 99251362 - 99390732 on Build GRCh38
- UniGene ID
- Hs.578684
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
W05392.1 |
- |
|
68 |
|
Chr. - - [-] |
GenBankmRNA |
CR593703.1 |
- |
1 |
1167 |
|
Chr. 15 - 99387611 - 99389069 [+] |
GenBankmRNA |
BE782162.1 |
- |
|
409 |
|
Chr. - - [-] |
GenBankmRNA |
BX110980.1 |
- |
1 |
270 |
|
Chr. 15 - 99388508 - 99389203 [+] |
View Full Product Details