Gene Transcripts
- Gene Symbol
- LOC101928517
- Entrez Gene ID
- 101928517
- Gene Name
- uncharacterized LOC101928517
- Gene Aliases
- Chromosome Location
- Chr. 19: 51152923 - 51181966 on Build GRCh38
- UniGene ID
- Hs.707693
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_110732.1 |
|
5 |
877 |
|
GenBank mRNA |
BC045766.1 |
- |
5 |
877 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000600074.1 |
- |
5 |
877 |
|
Chr. 19 - 51152923 - 51181966 [-] |
View Full Product Details