Gene Transcripts

Gene Symbol
PLXDC2
Entrez Gene ID
84898
Gene Name
plexin domain containing 2
Gene Aliases
TEM7R
Chromosome Location
Chr. 10: 19816215 - 20289856 on Build GRCh38
UniGene ID
Hs.658134
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282736.1 NP_001269665.1 13 5042
NM_032812.8 NP_116201.7 14 5189
GenBank mRNA AK127644.1 - 1 1766

View Full Product Details