Gene Transcripts
- Gene Symbol
- LINC01231
- Entrez Gene ID
- 101929247
- Gene Name
- long intergenic non-protein coding RNA 1231
- Gene Aliases
- Chromosome Location
- Chr. 9: 3181589 - 3200500 on Build GRCh38
- UniGene ID
- Hs.547178
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_121585.1 |
|
4 |
648 |
|
GenBank mRNA |
BC041456.1 |
- |
4 |
648 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BI756497.1 |
- |
4 |
648 |
|
Chr. 9 - 3181589 - 3198525 [+] |
View Full Product Details