Gene Transcripts
- Gene Symbol
- MOB3B
- Entrez Gene ID
- 79817
- Gene Name
- MOB kinase activator 3B
- Gene Aliases
- C9orf35, MOB1D, MOBKL2B
- Chromosome Location
- Chr. 9: 27325209 - 27529852 on Build GRCh38
- UniGene ID
- Hs.369022
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK022201.1 |
- |
1 |
110 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AU121522.1 |
- |
1 |
110 |
|
Chr. 9 - 27336933 - 27337784 [-] |
View Full Product Details