Gene Transcripts
- Gene Symbol
- NDE1
- Entrez Gene ID
- 54820
- Gene Name
- nudE neurodevelopment protein 1
- Gene Aliases
- HOM-TES-87, LIS4, MHAC, NDE, NUDE, NUDE1
- Chromosome Location
- Chr. 16: 15643267 - 15726353 on Build GRCh38
- UniGene ID
- Hs.655378
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BM719917.1 |
- |
1 |
73 |
|
Chr. 16 - 15699638 - 15700159 [+] |
GenBankmRNA |
BE714767.1 |
- |
2 |
265 |
|
Chr. 16 - 15696728 - 15699748 [+] |
GenBankmRNA |
BU160687.1 |
- |
5 |
634 |
|
Chr. 16 - 15687427 - 15699914 [+] |
GenBankmRNA |
BF749885.1 |
- |
2 |
287 |
|
Chr. 16 - 15696703 - 15699798 [+] |
GenBankmRNA |
BF748306.1 |
- |
1 |
64 |
|
Chr. 16 - 15699655 - 15699894 [+] |
View Full Product Details