Gene Transcripts
- Gene Symbol
- FAM110C
- Entrez Gene ID
- 642273
- Gene Name
- family with sequence similarity 110 member C
- Gene Aliases
- Chromosome Location
- Chr. 2: 38814 - 47077 on Build GRCh38
- UniGene ID
- Hs.8379
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AL137761.1 |
- |
1 |
155 |
|
View Full Product Details