Gene Transcripts

Gene Symbol
THEGL
Entrez Gene ID
100506564
Gene Name
theg spermatid protein-like
Gene Aliases
Chromosome Location
Chr. 4: 56530575 - 56626214 on Build GRCh38
UniGene ID
Hs.573073
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011534357.1 XP_011532659.1 11 1807
GenBank mRNA AL833514.1 - 2 3223

View Full Product Details