Gene Transcripts
- Gene Symbol
- THEGL
- Entrez Gene ID
- 100506564
- Gene Name
- theg spermatid protein-like
- Gene Aliases
- Chromosome Location
- Chr. 4: 56530575 - 56626214 on Build GRCh38
- UniGene ID
- Hs.573073
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AL833514.1 |
- |
1 |
2344 |
|
View Full Product Details