Gene Transcripts
- Gene Symbol
- LINC00560
- Entrez Gene ID
- 100861553
- Gene Name
- long intergenic non-protein coding RNA 560
- Gene Aliases
- Chromosome Location
- Chr. 13: 88626048 - 88630397 on Build GRCh38
- UniGene ID
- Hs.560132
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC043278.2 |
- |
1 |
358 |
|
View Full Product Details