Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001013438.2 | NP_001013456.1 | 16 | 8177 | |
NM_001013439.2 | NP_001013457.1 | 18 | 8613 | ||
NM_005087.3 | NP_005078.2 | 17 | 8269 | ||
GenBank mRNA | AF075028.1 | - | 1 | 264 |