Gene Transcripts
- Gene Symbol
- LOC100131150
- Entrez Gene ID
- 100131150
- Gene Name
- uncharacterized LOC100131150
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.124147
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK096194.1 |
- |
1 |
345 |
|
- Gene Symbol
- KDM2A
- Entrez Gene ID
- 22992
- Gene Name
- lysine demethylase 2A
- Gene Aliases
- CXXC8, FBL11, FBL7, FBXL11, JHDM1A, LILINA
- Chromosome Location
- Chr. 11: 67119210 - 67258082 on Build GRCh38
- UniGene ID
- Hs.124147
- Species
- Homo sapiens
View Full Product Details