Gene Transcripts
- Gene Symbol
- SNRPD2P2
- Entrez Gene ID
- 645339
- Gene Name
- small nuclear ribonucleoprotein D2 pseudogene 2
- Gene Aliases
- Chromosome Location
- Chr. 1: 231475762 - 231476525 on Build GRCh38
- UniGene ID
- Hs.382049
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_033826.1 |
|
1 |
393 |
|
GenBank mRNA |
BC014600.1 |
- |
1 |
399 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BG722592.1 |
- |
1 |
399 |
|
Chr. 1 - 231475762 - 231476528 [-] |
View Full Product Details