Gene Transcripts
- Gene Symbol
- OPA1
- Entrez Gene ID
- 4976
- Gene Name
- OPA1, mitochondrial dynamin like GTPase
- Gene Aliases
- BERHS, MGM1, MTDPS14, NPG, NTG, largeG
- Chromosome Location
- Chr. 3: 193593144 - 193697811 on Build GRCh38
- UniGene ID
- Hs.594504
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK022107.1 |
- |
1 |
970 |
|
View Full Product Details