Gene Transcripts

Gene Symbol
TNRC6C
Entrez Gene ID
57690
Gene Name
trinucleotide repeat containing 6C
Gene Aliases
Chromosome Location
Chr. 17: 77974984 - 78108835 on Build GRCh38
UniGene ID
Hs.406810
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006721996.3 XP_006722059.3 1 4223
XM_006721997.3 XP_006722060.3 1 4223
XM_006722000.3 XP_006722063.3 1 4223
XM_017024906.1 XP_016880395.1 1 4223
XM_017024907.1 XP_016880396.1 1 4223
XM_017024908.1 XP_016880397.1 1 4223
GenBank mRNA BC035105.1 - 1 3117
BC051468.1 - 1 3112

View Full Product Details