Gene Transcripts

Gene Symbol
CERS5
Entrez Gene ID
91012
Gene Name
ceramide synthase 5
Gene Aliases
LASS5, Trh4
Chromosome Location
Chr. 12: 50129306 - 50167533 on Build GRCh38
UniGene ID
Hs.270525
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK055740.1 - 1 610

View Full Product Details