Gene Transcripts
- Gene Symbol
- PRR29-AS1
- Entrez Gene ID
- 400612
- Gene Name
- PRR29 antisense RNA 1
- Gene Aliases
- Chromosome Location
- Chr. 17: 63996071 - 63999899 on Build GRCh38
- UniGene ID
- Hs.591170
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_126169.1 |
|
3 |
2596 |
|
GenBank mRNA |
AK094767.1 |
- |
3 |
2596 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000580942.1 |
- |
3 |
2596 |
|
Chr. 17 - 63996071 - 63999899 [-] |
View Full Product Details