Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001039960.2 | NP_001035049.1 | 25 | 7299 | |
NM_001258401.2 | NP_001245330.1 | 25 | 7245 | ||
XM_011539010.1 | XP_011537312.1 | 26 | 7354 | ||
XM_011539011.1 | XP_011537313.1 | 26 | 7299 | ||
XM_011539014.2 | XP_011537316.1 | 16 | 5952 | ||
XM_017020240.1 | XP_016875729.1 | 26 | 7381 | ||
XM_017020242.1 | XP_016875731.1 | 26 | 7368 | ||
GenBank mRNA | AK124576.1 | - | 1 | 497 | |
EU637925.1 | - | 1 | 3106 |