Gene Transcripts
- Gene Symbol
- FLJ38773
- Entrez Gene ID
- 284808
- Gene Name
- uncharacterized protein FLJ38773
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK096092.1 |
- |
1 |
1253 |
|
View Full Product Details