Gene Transcripts
- Gene Symbol
- BTG3
- Entrez Gene ID
- 10950
- Gene Name
- BTG anti-proliferation factor 3
- Gene Aliases
- ANA, APRO4, TOB5, TOB55, TOFA
- Chromosome Location
- Chr. 21: 17593650 - 17612950 on Build GRCh38
- UniGene ID
- Hs.473420
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC028229.1 |
- |
1 |
2119 |
|
View Full Product Details