Gene Transcripts
- Gene Symbol
- ZNF827
- Entrez Gene ID
- 152485
- Gene Name
- zinc finger protein 827
- Gene Aliases
- Chromosome Location
- Chr. 4: 145757627 - 145939008 on Build GRCh38
- UniGene ID
- Hs.133916
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BF965409.1 |
- |
|
232 |
|
Chr. - - [-] |
GenBankmRNA |
BU536382.1 |
- |
1 |
158 |
|
Chr. 4 - 145938611 - 145938842 [-] |
GenBankmRNA |
HY165912.1 |
- |
1 |
133 |
|
Chr. 4 - 145938502 - 145938820 [-] |
View Full Product Details