Gene Transcripts

Gene Symbol
SPTBN2
Entrez Gene ID
6712
Gene Name
spectrin beta, non-erythrocytic 2
Gene Aliases
GTRAP41, SCA5, SCAR14
Chromosome Location
Chr. 11: 66682496 - 66729361 on Build GRCh38
UniGene ID
Hs.26915
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_005274192.4 XP_005274249.1 38 10235
XM_005274193.3 XP_005274250.1 37 10009
XM_006718669.3 XP_006718732.1 36 11922
XM_006718671.3 XP_006718734.1 38 9635
XM_017018174.1 XP_016873663.1 38 10327
XM_017018175.1 XP_016873664.1 38 9956
XM_017018176.1 XP_016873665.1 38 9676
XM_017018177.1 XP_016873666.1 37 9865
XM_017018178.1 XP_016873667.1 37 9548
GenBank mRNA AK021479.1 - 1 1721


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BF351671.1 - 1 295 Chr. 11 - 66683363 - 66683829 [-]

View Full Product Details