Gene Transcripts
- Gene Symbol
- FSBP
- Entrez Gene ID
- 100861412
- Gene Name
- fibrinogen silencer binding protein
- Gene Aliases
- Chromosome Location
- Chr. 8: 94427712 - 94436952 on Build GRCh38
- UniGene ID
- Hs.30561
- Species
- Homo sapiens
- Gene Symbol
- RAD54B
- Entrez Gene ID
- 25788
- Gene Name
- RAD54 homolog B (S. cerevisiae)
- Gene Aliases
- RDH54
- Chromosome Location
- Chr. 8: 94371960 - 94475115 on Build GRCh38
- UniGene ID
- Hs.30561
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BE887706.1 |
- |
|
322 |
|
Chr. - - [-] |
View Full Product Details