Gene Transcripts
- Gene Symbol
- KLF8
- Entrez Gene ID
- 11279
- Gene Name
- Kruppel like factor 8
- Gene Aliases
- BKLF3, ZNF741
- Chromosome Location
- Chr. X: 55908123 - 56291531 on Build GRCh38
- UniGene ID
- Hs.646614
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CD387915.1 |
- |
|
203 |
|
Chr. - - [-] |
GenBankmRNA |
CF130112.1 |
- |
1 |
24 |
|
Chr. X - 55908381 - 56015147 [+] |
View Full Product Details