Gene Transcripts
- Gene Symbol
- CLLU1
- Entrez Gene ID
- 574028
- Gene Name
- chronic lymphocytic leukemia up-regulated 1
- Gene Aliases
- Chromosome Location
- Chr. 12: 92421531 - 92431002 on Build GRCh38
- UniGene ID
- Hs.730377
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AJ845167.1 |
- |
1 |
1997 |
|
View Full Product Details