Gene Transcripts

Gene Symbol
ITGBL1
Entrez Gene ID
9358
Gene Name
integrin subunit beta like 1
Gene Aliases
OSCP, TIED
Chromosome Location
Chr. 13: 101452593 - 101720857 on Build GRCh38
UniGene ID
Hs.696554
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271754.1 NP_001258683.1 11 3927
XM_005254100.4 XP_005254157.1 12 4103
GenBank mRNA AK026784.1 - 1 948
AL527690.3 - 1 86


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA DB205402.1 - 1 384 Chr. 13 - 101719518 - 101720086 [+]

View Full Product Details