Gene Transcripts
- Gene Symbol
- LOC105377468
- Entrez Gene ID
- 105377468
- Gene Name
- uncharacterized LOC105377468
- Gene Aliases
- Chromosome Location
- Chr. 4: 146052386 - 146059057 on Build GRCh38
- UniGene ID
- Hs.148809
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AL832220.1 |
- |
1 |
1222 |
|
View Full Product Details