Gene Transcripts
- Gene Symbol
- FUBP3
- Entrez Gene ID
- 8939
- Gene Name
- far upstream element binding protein 3
- Gene Aliases
- FBP3
- Chromosome Location
- Chr. 9: 130579567 - 130638352 on Build GRCh38
- UniGene ID
- Hs.673029
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC015184.1 |
- |
1 |
820 |
|
View Full Product Details