Gene Transcripts
- Gene Symbol
- CMTM6
- Entrez Gene ID
- 54918
- Gene Name
- CKLF like MARVEL transmembrane domain containing 6
- Gene Aliases
- CKLFSF6, PRO2219
- Chromosome Location
- Chr. 3: 32481312 - 32502911 on Build GRCh38
- UniGene ID
- Hs.380627
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC042995.1 |
- |
|
145 |
|
View Full Product Details