Gene Transcripts

Gene Symbol
PEX11A
Entrez Gene ID
8800
Gene Name
peroxisomal biogenesis factor 11 alpha
Gene Aliases
PEX11-ALPHA, PMP28, hsPEX11p
Chromosome Location
Chr. 15: 89681531 - 89690784 on Build GRCh38
UniGene ID
Hs.31034
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271572.1 NP_001258501.1 3 1140
NM_001271573.1 NP_001258502.1 2 1117
NM_003847.2 NP_003838.1 3 1233
GenBank mRNA AK297442.1 - 2 965
AL360141.1 - 1 347

View Full Product Details