Gene Transcripts
- Gene Symbol
- ZZEF1
- Entrez Gene ID
- 23140
- Gene Name
- zinc finger ZZ-type and EF-hand domain containing 1
- Gene Aliases
- ZZZ4
- Chromosome Location
- Chr. 17: 4004445 - 4143020 on Build GRCh38
- UniGene ID
- Hs.277624
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK001683.1 |
- |
15 |
2640 |
|
BC047285.1 |
- |
15 |
2803 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
U33270.1 |
- |
1 |
49 |
|
Chr. 17 - 4021952 - 4022483 [-] |
View Full Product Details