Gene Transcripts
- Gene Symbol
- D21S2088E
- Entrez Gene ID
- 266917
- Gene Name
- D21S2088E
- Gene Aliases
- Chromosome Location
- Chr. 21: 23361104 - 23384835 on Build GRCh38
- UniGene ID
- Hs.145622
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_040254.1 |
|
2, 3 |
287 |
|
GenBank mRNA |
AY063451.1 |
- |
2, 3 |
287 |
|
View Full Product Details