Gene Transcripts
- Gene Symbol
- LINC00559
- Entrez Gene ID
- 100874187
- Gene Name
- long intergenic non-protein coding RNA 559
- Gene Aliases
- Chromosome Location
- Chr. 13: 90060247 - 90119717 on Build GRCh38
- UniGene ID
- Hs.564526
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_047489.1 |
|
2 |
1401 |
|
GenBank mRNA |
BC043442.1 |
- |
2 |
1405 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000569476.2 |
- |
2 |
1403 |
|
Chr. 13 - 90060252 - 90119719 [-] |
View Full Product Details