Gene Transcripts
- Gene Symbol
- DPCD
- Entrez Gene ID
- 25911
- Gene Name
- deleted in primary ciliary dyskinesia homolog (mouse)
- Gene Aliases
- Chromosome Location
- Chr. 10: 101588332 - 101609653 on Build GRCh38
- UniGene ID
- Hs.658128
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK296220.1 |
- |
4 |
1170 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AK130871.1 |
- |
3 |
398 |
|
Chr. 10 - 101587215 - 101588879 [+] |
View Full Product Details