Gene Transcripts
- Gene Symbol
- FLJ22447
- Entrez Gene ID
- 400221
- Gene Name
- uncharacterized LOC400221
- Gene Aliases
- Chromosome Location
- Chr. 14: 61570540 - 61654713 on Build GRCh38
- UniGene ID
- Hs.344000
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_039985.1 |
|
1 |
94 |
|
GenBank mRNA |
AK026100.1 |
- |
1 |
98 |
|
BQ439352.1 |
- |
1 |
183 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BX339593.2 |
- |
1 |
38 |
|
Chr. 14 - 61570596 - 61653843 [+] |
View Full Product Details