Gene Transcripts

Gene Symbol
C6
Entrez Gene ID
729
Gene Name
complement component 6
Gene Aliases
Chromosome Location
Chr. 5: 41142116 - 41261486 on Build GRCh38
UniGene ID
Hs.481992
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA J05063.1 - 1 376


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BX502917.1 - 1 229 Chr. 5 - 41159261 - 41159994 [-]

View Full Product Details