Gene Transcripts

Gene Symbol
STPG2
Entrez Gene ID
285555
Gene Name
sperm tail PG-rich repeat containing 2
Gene Aliases
C4orf37
Chromosome Location
Chr. 4: 97441450 - 98143473 on Build GRCh38
UniGene ID
Hs.680226
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BX648305.1 - 5 2277


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BC041952.1 - 1 1029 Chr. 7 - 80421513 - 80423208 [+]

View Full Product Details