Gene Transcripts
- Gene Symbol
- LOC101926942
- Entrez Gene ID
- 101926942
- Gene Name
- uncharacterized LOC101926942
- Gene Aliases
- Chromosome Location
- Chr. 10: 90402521 - 90540805 on Build GRCh38
- UniGene ID
- Hs.382666
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_110657.1 |
|
7 |
943 |
|
GenBank mRNA |
BC030278.1 |
- |
7 |
943 |
|
View Full Product Details