Gene Transcripts

Gene Symbol
FAM63B
Entrez Gene ID
54629
Gene Name
family with sequence similarity 63 member B
Gene Aliases
Chromosome Location
Chr. 15: 58771192 - 58861900 on Build GRCh38
UniGene ID
Hs.591122
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040450.2 NP_001035540.1 9 7009
NM_001040453.2 NP_001035543.1 9 6836
XM_011521687.1 XP_011519989.1 10 7061
XM_011521688.1 XP_011519990.1 10 7058
XM_011521689.1 XP_011519991.1 10 6959
XM_011521690.1 XP_011519992.1 9 6905
XM_017022332.1 XP_016877821.1 8 6815
XM_017022333.1 XP_016877822.1 7 6761
XM_017022334.1 XP_016877823.1 7 6758
GenBank mRNA AK021710.1 - 1 1079

View Full Product Details