Gene Transcripts
- Gene Symbol
- LOC100291666
- Entrez Gene ID
- 100291666
- Gene Name
- serologically defined breast cancer antigen NY-BR-40
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.736534
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF308290.1 |
- |
1 |
350 |
|
View Full Product Details