Gene Transcripts
- Gene Symbol
- HOTTIP
- Entrez Gene ID
- 100316868
- Gene Name
- HOXA distal transcript antisense RNA
- Gene Aliases
- HOXA-AS6, HOXA13-AS1, NCRNA00213
- Chromosome Location
- Chr. 7: 27200421 - 27207259 on Build GRCh38
- UniGene ID
- Hs.199486
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_037843.3 |
|
2 |
1007 |
|
GenBank mRNA |
AK093987.1 |
- |
2 |
991 |
|
GU724873.1 |
- |
2 |
1003 |
|
View Full Product Details