Gene Transcripts

Gene Symbol
HOTTIP
Entrez Gene ID
100316868
Gene Name
HOXA distal transcript antisense RNA
Gene Aliases
HOXA-AS6, HOXA13-AS1, NCRNA00213
Chromosome Location
Chr. 7: 27200421 - 27207259 on Build GRCh38
UniGene ID
Hs.199486
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_037843.3 2 335
GenBank mRNA AK093987.1 - 2 319
GU724873.1 - 2 335


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA DA858617.1 - 2 301 Chr. 7 - 27200456 - 27201153 [+]
GenBankmRNA DB250371.1 - 2 319 Chr. 7 - 27200437 - 27201097 [+]

View Full Product Details